Learn More
3M™ Spiral Hoses
Product Code. 11395262 Shop All 3M Products
Change view
Click to view available options
Length (Metric):
7.5 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11395262 7.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11395262 Supplier 3M™ Supplier No. 7000042678
  • ONLINE EXCLUSIVE PRICE
    341.00€
    232.72€ / 1 pièces
    Économisez 108.28€
    31% de réduction
 Restricted Item
Expédition estimée: 28-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Spiral Hose
For Use With (Application) Compressed Air
Quantity 1 Ea.
Length (Metric) 7.5 m