Learn More
Thermo Scientific™ SPME Arrow Liner
Product Code. 17644145 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
ID 1.3 mm
ID 1.7 mm x 6.5 mm x 78.5 mm
Quantity:
3 Pk
3 Pk.
5 Pk.
Unit Size:
Pack of 3
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Unit Size
17644145 ID 1.7 mm x 6.5 mm x 78.5 mm 5 Pk. Pack of 5
15840705 ID 1.3 mm 3 Pk. Pack of 3
15850705 ID 1.7 mm x 6.5 mm x 78.5 mm 3 Pk Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17644145 Supplier Thermo Scientific™ Supplier No. 453A0415UI
Expédition estimée: 11-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type SPME Arrow Liner
For Use With (Equipment) Triplus RSH Autosampler, TRACE 1300 Series Gas Chromatograph and GC-MS Systems
Description ID 1.7 mm x 6.5 mm x 78.5 mm
Quantity 5 Pk.