Learn More
Thermo Scientific™ SPME Arrow and SPME Fibers for Solid Phase Micro Extraction
Product Code. 18875425 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Dark Gray
Material:
DVB/Carbon WR/PDMS
Diameter (Metric) Outer:
23 Gauge
Quantity:
3/Pk.
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material Diameter (Metric) Outer Quantity Unit Size
18875425 Dark Gray DVB/Carbon WR/PDMS 23 Gauge 3/Pk. Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18875425 Supplier Thermo Scientific™ Supplier No. 36SP05T3
 Restricted Item
En stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Automate solvent-free solid phase micro extraction (SPME) sample preparation for GC and GC/MS by performing sample extraction and enrichment in a single step.
Automate solvent-free solid phase micro extraction (SPME) sample preparation by performing sample extraction and enrichment in a single step using Thermo Scientific™ SPME Arrow and SPME Fiber products. These products are used with the Thermo Scientific™ TriPlus RSH™ autosampler prior to gas chromatography (GC) or gas chromatography mass spectrometry (GC/MS) analysis. The SPME Arrow and Fiber products are designed for volatile and semi-volatile compounds having a large range of boiling point and polarity. This sample preparation technique is ideal for ultra-trace and trace GC and GC/MS analysis of solid, liquid, or gas samples from environmental, food, clinical, and forensic industries.
SPME Fiber and Arrow products
The SPME Arrow and Fiber comprise a thin fused-silica fiber coated by an organic phase. They extract and concentrate analytes by ab- or adsorption and desorption processes. The fiber is successively thermally desorbed into a GC injector and the analytes are transferred to the analytical column. Selecting SPME Fiber or SPME Arrow depends on hardware compatibility and which product is used in your validated laboratory method.
SPME Fiber
  • Original proven design used by many customers in standard methods
  • Wide selection of sorption materials
  • Requires SPME Fiber Conditioning Module
SPME Arrow
  • Improved, optimized fiber geometry
  • Faster extraction, higher throughput, more robust
  • Wide selection of sorption materials
  • Requires compatible firmware, SPME Arrow Conditioning Module and Heatex Stirrer, and an adaptation kit for the GC inlet of the TRACE1300 Series GC Instant Connect Split Splitless injector to accommodate the larger diameter of SPME Arrows
GC Compatibility
SPME Arrow and SPME Fiber products are for use with the TriPlus RSH autosampler with GC analysis using the Thermo Scientific TRACE 1300 Series GC with an Instant Connect Split/Splitless injector or a TRACE GC Ultra Split/Splitless injector. Please select the Support button near the top of this page to ask Thermo Fisher Scientific representative about compatibility with other GC autosampler manufacturers.
TRUSTED_SUSTAINABILITY
Specifications
Thickness (Metric) 50/30 μm
Length (Metric) 10 mm
Material DVB/Carbon WR/PDMS
Quantity 3/Pk.
Product Type Fiber
Phase DVB/Carbon WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane)
Color Dark Gray
Diameter (Metric) Outer 23 Gauge
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.