Learn More
Hach Lange™ StablCal™ Calibration Set
Product Code. 10763612
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10763612 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10763612 Supplier Hach Lange™ Supplier No. 2662110
  • ONLINE EXCLUSIVE PRICE
    634.71€
    513.00€ / 1 pièces
    Économisez 121.71€
    19% de réduction
 Restricted Item
Expédition estimée: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type StablCal Calibration Set
Volume (Metric) 100 mL
compliance-icons
Product Identifier
  • Formazin Turbidity Standard 4000 NTU
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P285-In case of inadequate ventilation wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P341-IF INHALED: If breathing is difficult, remove victim to fresh air and keep at rest in a position comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P501-Dispose of contents/container to .