Learn More
Kartell SpA Stackable Deep Tray
Product Code. 12081103
Change view
Click to view available options
Capacity (Metric):
10 L
16 L
20 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12081103 20 L Each
12071103 10 L Each
11381105 16 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12081103 Supplier Kartell SpA Supplier No. 0054400
  • ONLINE EXCLUSIVE PRICE
    34.00€
    31.92€ / 1 pièces
    Économisez 2.08€
    6% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Deep tray, low form, stackable
Stackable deep trays made of HDPE with handles
Specifications
Capacity (Metric) 20 L
Closure Type None
Color White
Material HDPE
Shape Rectangular
Product Type Deep Tray
Sterility Non-sterile
Dimensions (L x W x H) 470 x 410 x 145 mm
Length (Metric) 470 mm
Width (Metric) 410 mm
Height (Metric) 145 mm