Learn More
Scotsman™ Stainless Steel AF103 AS Ice Flaker
Product Code. 11764798
Change view
Click to view available options
Capacity (Metric):
30 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11764798 30 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11764798 Supplier Scotsman™ Supplier No. AF107AS
  • ONLINE EXCLUSIVE PRICE
    13470.61€
    5078.64€ / 1 pièces
    Économisez 8391.97€
    62% de réduction
 Restricted Item
Expédition estimée: 23-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Specifications
Depth (English) 24.5 in.
Capacity (English) 66 lb.
Capacity (Metric) 30 kg
Depth (Metric) 622 mm
For Use With (Application) Flake ice production
Width (English) 23.31 in.
Width (Metric) 592 mm
Yield Up to 108 kg (238 lb.) of flake ice per day