Learn More
Dueperthal™ Stainless Steel Can Collector
Product Code. 10490961
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10490961 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10490961 Supplier Dueperthal™ Supplier No. 5.10.006
 Restricted Item
Expédition estimée: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Capacity (Metric) 6 L
Diameter (Metric) Outer 250 mm
Color Blue
Finish Stove enamelled
Height (Metric) 200 mm
For Use With (Application) Use as safety containers or cleaning containers
Material Stainless Steel