Learn More
De Buyer™ Stainless Steel Flat Bottom Pastry Bowl
Product Code. 11930605
Change view
Click to view available options
Capacity (Metric):
16 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11930605 16 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11930605 Supplier De Buyer™ Supplier No. 3250.40
  • ONLINE EXCLUSIVE PRICE
    212.03€
    61.20€ / 1 pièces
    Économisez 150.83€
    71% de réduction
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A multifunctional bowl with an open rolled rim and no sharp edges so that the entire mixture can be beaten.
  • Used for making mixtures, whisking, heating by bain-marie, and more
  • Can go into the refrigerator, deep freezer, blast-chiller and oven
  • All stainless steel construction, robust and resistant to deformation
  • Wide, flat bottom enables greater stability
  • Functional and ergonomic open rolled rim which enables a secure, strong and comfortable grip for dynamic whisking
  • Dishwasher-safe
Specifications
Material Stainless Steel
Capacity (Metric) 16 L
Height (Metric) 17.5 cm
Diameter (Metric) 40 cm
Shape Conical, Flat Bottom
Product Type Pastry Bowl