Learn More
Retsch™ Stainless Steel Knife
Product Code. 15860530 Shop All Retsch Products
Change view
Click to view available options
Product Type:
Metal Blade and Knife Cylinder
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15860530 Metal Blade and Knife Cylinder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15860530 Supplier Retsch™ Supplier No. 02.446.0049
  • ONLINE EXCLUSIVE PRICE
    1176.00€
    909.00€ / 1 pièces
    Économisez 267.00€
    22% de réduction
Expédition estimée: 18-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Retsch™ Stainless Steel Knife is designed for use with Knife Mill GRINDOMIX GM 300.
  • Constructed of stainless steel
Specifications
Product Type Metal Blade and Knife Cylinder
For Use With (Equipment) Knife Mill GRINDOMIX GM 300
Material Stainless Steel