Learn More
Bochem™ Stainless Steel Laboratory Carts 1
Product Code. 10084220
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10084220 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10084220 Supplier Bochem™ Supplier No. 11411
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Laboratory Cart
Dimensions (L x W x H) 900 x 600 x 940 mm (with wheel and handle)
Height (Metric) 940 mm
Length (Metric) 900 mm
Load Bearing Capacity (Metric) 120 kg
Material Stainless Steel
No. of Shelves 3
Width (Metric) 600 mm