Learn More
DE Buyer™ Stainless Steel Measuring Jug
Product Code. 11923045
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11923045 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11923045 Supplier DE Buyer™ Supplier No. 3565.025N
  • ONLINE EXCLUSIVE PRICE
    37.01€
    11.70€ / 1 pièces
    Économisez 25.31€
    68% de réduction
Expédition estimée: 07-12-2026