Learn More
- Home
- Products
- Balances and Scales
- Calibration Weights
- Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight Without DAkkS Calibration Certificate
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight Without DAkkS Calibration Certificate
Product Code. 10216131 Shop All Sartorius Products
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
50 kg
Unit Size:
Each
24 product options available for selection
| Product Code. | Mass | Unit Size |
|---|---|---|
| 10216131 | 100 g | Each |
| 10022201 | 100 mg | Each |
| 10554751 | 10 kg | Each |
| 10684322 | 10 mg | Each |
| 10461612 | 10 g | Each |
| 10022121 | 1 kg | Each |
| 10347301 | 1 g | Each |
| 10767422 | 1 mg | Each |
| 10440076 | 200 g | Each |
| 10642022 | 20 g | Each |
| 10594181 | 20 mg | Each |
| 10225891 | 200 mg | Each |
| 10114131 | 20 kg | Each |
| 10317591 | 2 g | Each |
| 10193691 | 2 mg | Each |
| 10595311 | 2 kg | Each |
| 10072091 | 500 g | Each |
| 10205941 | 500 mg | Each |
| 10777422 | 50 kg | Each |
| 10622032 | 50 g | Each |
| 10632022 | 50 mg | Each |
| 10265841 | 5 g | Each |
| 10692022 | 5 mg | Each |
| 10480842 | 5 kg | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
This item is not returnable.
View return policy
Product Code.
10216131
Supplier
Sartorius™
Supplier No.
YCW51100
You must Sign In and have a business account to purchase this item.
Restricted Item
Expédition estimée: 22-01-2027
Connectez-vous pour voir le stock disponible
Connectez-vous pour voir le stock disponible
Add to basket
Add to basket
This item is not returnable.
View return policy