Learn More
AMSTA Rostfritt Stainless Steel Trolley with 4 Castors
Product Code. 11711786
Change view
Click to view available options
Diameter (Metric):
350 mm
400 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11711786 400 mm Each
11711776 350 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11711786 Supplier AMSTA Rostfritt Supplier No. 5070850
  • ONLINE EXCLUSIVE PRICE
    317.63€
    249.16€ / 1 pièces
    Économisez 68.47€
    21% de réduction
Expédition estimée: 18-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-quality stainless steel trolley, available in various sizes.
  • Quality
  • Invisible weld seams
  • Pharmaceutical grade finish
  • Sustainable production
  • Made in Sweden
  • Two castors with brakes and two castors without
Specifications
Type Drum Dolly
Holds 1 Drum (20 to 55 gal.)
Load Bearing Capacity (English) 264.6 lb.
Load Bearing Capacity (Metric) 120 kg
Material Steel
Color Silver
Diameter (Metric) 400 mm