Learn More
HUBER Stainless Steel Unipump I DC
Product Code. 10783856
Change view
Click to view available options
Maximum Pressure:
1.0 bar
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Maximum Pressure Unit Size
10783856 1.0 bar Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10783856 Supplier HUBER Supplier No. 1085.0001.00
  • ONLINE EXCLUSIVE PRICE
    3805.00€
    3210.00€ / 1 pièces
    Économisez 595.00€
    15% de réduction
Expédition estimée: 10-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Features external pressure pump.
  • Bearing out of liquid circuit
  • All parts in liquid circuit and housing made of stainless steel
  • Operation either manual (on/off) or external via 3-line-connector
  • Cable (Option) for the potentional free contact of Unistat Tango and the big power Unistats
  • Compatible with thermostats and unichillers
Specifications
Overall Height (Metric) 214 mm
Overall Width (Metric) 219 mm
Temperature (Metric) Max -120°C to +300°C
Maximum Pressure 1.0 bar