Learn More
WTW™ Standard Calibration and Storage Sleeve for WTW™ Oxygen Meter
Product Code. 10519911
Change view
Click to view available options
For Use With (Equipment):
Oxygen meter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10519911 Oxygen meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10519911 Supplier WTW™ Supplier No. 205362
Expédition estimée: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The calibration and storage containers for the WTW oxygen sensors are suitable for the standard conform calibration (DIN ISO 5814 or DIN ISO 17289: 2014-12) with water steam saturated air as well as for storage of the sensors to protect the sensitive membrane.
TRUSTED_SUSTAINABILITY
Specifications
Description Standard calibration and storage sleeve
For Use With (Equipment) Oxygen meter