Learn More
3M™ Standard Duty Compressed Air Supply Tubing
Product Code. 12354679 Shop All 3M Products
Change view
Click to view available options
Length (Metric):
10 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12354679 10 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12354679 Supplier 3M™ Supplier No. 7000044242
  • ONLINE EXCLUSIVE PRICE
    624.00€
    429.07€ / 1 pièces
    Économisez 194.93€
    31% de réduction
 Restricted Item
Expédition estimée: 18-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Respirator
For Use With (Application) Respirator system
Quantity 1 Ea.
Length (Metric) 10 m