Learn More
STEMCELL Technologies STEMdiff™ NK Cell Kit
Product Code. 17177491
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17177491 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177491 Supplier STEMCELL Technologies Supplier No. 1000170
 Restricted Item
Expédition estimée: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For expansion and differentiation of hPSCs to NK cells
  • STEMdiff™ NK Cell Kit includes serum-free media and supplements for the feeder-free differentiation of human pluripotent stem cells (PSCs) to natural killer (NK) cells expressing CD56
  • STEMdiff™ Hematopoietic - EB reagents are animal-component free and optimized for lymphoid differentiation potential
  • Using these reagents, CD34+ hematopoietic progenitor cells are first generated from PSCs; these can then be differentiated to NK cells using the StemSpan™ components
Specifications
Cell Type Hematopoietic Cells, PSC-Derived, NK Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation, Expansion
Quantity 1 Kit