Learn More
STEMCELL Technologies StemSpan™ NK Cell Generation Kit
Product Code. 17199961
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17199961 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17199961 Supplier STEMCELL Technologies Supplier No. 09960
 Restricted Item
Expédition estimée: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For expansion and differentiation of human CD34+ hematopoietic progenitor cells to NK cells
  • StemSpan™ NK Cell Generation Kit has been developed to differentiate CD34+ cells isolated from cord blood (CB) and bone marrow (BM) to natural killer (NK) cells, without use of stromal cells
  • This kit typically promotes the expansion of thousands of CD56+ NK cells in cultures initiated with CD34+ human CB cells
  • StemSpan™ Lymphoid Progenitor Expansion Supplement (10X) contains a combination of recombinant human cytokines and other additives formulated to selectively promote the expansion and differentiation of CD34+ cells isolated from human CB and BM samples to lymphoid progenitor cells when used in combination with StemSpan™ SFEM II medium, and on plates coated with StemSpan™ Lymphoid Differentiation Coating Material (100X)
  • Subsequently, StemSpan™ NK Cell Differentiation Supplement (100X) enables differentiation of lymphoid progenitor cells to NK cells
Specifications
Cell Type Hematopoietic Stem and Progenitor Cells, NK Cells
For Use With (Application) Cell Culture; Differentiation; Expansion
Quantity 1 Kit