Learn More
R&D Systems™ StemXVivo Adipogenic Supplement (100X)
Description
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into adipocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
- Contains high quality differentiation factors to drive reproducible and efficient MSC adipogenesis
- Is defined to reduce unwanted experimental variability
- Has been developed and optimized using MSCs
Spécification
Spécification
| Product Line | R&D Systems |
| Quantity | 3.5 mL |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |