Learn More
Aalborg™ Stainless Steel Stepping Motor Valve
Product Code. 12646173
Change view
Click to view available options
Type:
Stepping Motor Valver
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12646173 Stepping Motor Valver Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12646173 Supplier Aalborg™ Supplier No. SMV30SVE2
  • ONLINE EXCLUSIVE PRICE
    1233.75€
    893.52€ / 1 pièces
    Économisez 340.23€
    27% de réduction
 Restricted Item
Expédition estimée: 27-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Aalborg™ Stainless Steel Stepping Motor Valve is for high flow regulation of gases or liquids.
  • High precision two-way metering valves in aluminum
  • Unparalleled precision and resolution in controlling
  • Operate continuously without overheating
Specifications
Type Stepping Motor Valver
Airflow Rate 500 L/min.
Waterflow Rate 14.2L/min.