Learn More
Huenersdorf™ Storage bins
Product Code. 10413501
Change view
Click to view available options
Dimensions (L x W x H):
115 x 100 x 75 mm
230 x 145 x 125 mm
240 x 145 x 125 mm
335 x 200 x 150 mm
335 x 205 x 155 mm
485 x 310 x 180 mm
Color:
Blue
Green
Red
Unit Size:
Each
Pack of 12
Pack of 26
Pack of 8
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Color Unit Size
10413501 230 x 145 x 125 mm Blue Each
10670073 485 x 310 x 180 mm Red Pack of 8
15973142 335 x 205 x 155 mm Blue Pack of 12
10580482 485 x 310 x 180 mm Green -
16377268 335 x 200 x 150 mm Blue -
10500682 485 x 310 x 180 mm Blue -
17561965 240 x 145 x 125 mm Blue Pack of 26
10338572 115 x 100 x 75 mm Blue -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10413501 Supplier Huenersdorf™ Supplier No. 653300
Spotlight Offers
Use promo code 28038
View all promotions
En stock
Add to basket
Edge
Add to basket
Specifications
Type Storage Bin
Material Polystyrene
Color Blue
Shape Rectangular
Mounting Type Stackable
Capacity (Metric) 3.82 L, 436 kg
Certifications/Compliance Food Safe RAL
For Use With (Application) Food Safe
Dimensions (L x W x H) 230 x 145 x 125 mm
Length (Metric) 232 mm
Width (Metric) 148 mm
Height (Metric) 126 mm