Learn More
Sudan IV-d6, TRC
Product Code. 30425062
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425062 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30425062 Supplier Toronto Research Chemicals Supplier No. TRCS6889225MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sudan 4 D6
CAS 1014689-18-9
Molecular Formula C24 2H6 H14 N4 O
Quantity 5 mg
Synonym 2-Naphthalen-3,4,5,6,7,8-d6-ol','1-[2-[2-methyl-4-[2-(2-methylphenyl)diazenyl]phenyl]diazenyl]-, 1-[2-[2-Methyl-4-[2-(2-methylphenyl)diazenyl]phenyl]diazenyl]-2-naphthalen-3,4,5,6,7,8-d6-ol
InChI Formula InChI=1S/C24H20N4O/c1-16-7-3-6-10-21(16)26-25-19-12-13-22(17(2)15-19)27-28-24-20-9-5-4-8-18(20)11-14-23(24)29/h3-15,29H,1-2H3/i4D,5D,8D,9D,11D,14D
SMILES [2H]c1c([2H])c([2H])c2c(N=Nc3ccc(cc3C)N=Nc4ccccc4C)c(O)c([2H])c([2H])c2c1[2H]
IUPAC Name 3,4,5,6,7,8-hexadeuterio-1-[(E)-[2-methyl-4-[(E)-(2-methylphenyl)diazenyl]phenyl]diazenyl]naphthalen-2-ol
Molecular Weight (g/mol) 386.48
Formula Weight 386.2014