Learn More
Metro™ Super Adjustable Super Erecta™ Wire Shelf, Chrome Finish
Product Code. 15996841
Change view
Click to view available options
Width (English):
14 in.
18 in.
21 in.
24 in.
Length (English):
24 in.
30 in.
36 in.
42 in.
48 in.
54 in.
60 in.
72 in.
Unit Size:
Each
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Length (English) Unit Size
15996841 24 in. 42 in. Each
11765669 24 in. 60 in. Each
15946851 24 in. 54 in. Each
15878076 18 in. 24 in. -
15946821 18 in. 60 in. Each
15946841 24 in. 30 in. Each
15916811 18 in. 30 in. Each
15966811 18 in. 42 in. Each
15916821 18 in. 54 in. Each
15906841 21 in. 72 in. Each
15946831 21 in. 36 in. Each
15966831 21 in. 48 in. Each
11745669 24 in. 36 in. Each
11755669 24 in. 48 in. Each
15976821 18 in. 72 in. Each
15986831 21 in. 60 in. Each
15906831 21 in. 24 in. Each
15906801 14 in. 36 in. Each
11735669 18 in. 48 in. Each
11725669 18 in. 36 in. Each
15926841 24 in. 24 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
This item is not returnable. View return policy
Product Code. 15996841 Supplier Metro™ Supplier No. A2442NC
  • ONLINE EXCLUSIVE PRICE
    184.00€
    105.86€ / 1 pièces
    Économisez 78.14€
    42% de réduction
Expédition estimée: 25-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Includes
Each shelf shipped with one pack of four nonconductive sleeve/wedges.
Unique, open-wire design of chrome finish shelf permits many configurations, free circulation of air, and minimizes dust accumulation.
  • Each shelf provides approximately 76% open area for increased light penetration and improved visibility of stored items.
  • Can be loaded and unloaded easily from all sides.
  • Hard, durable surface.
  • Abrasion resistant.
  • Ideal for dry, noncorrosive environments.
  • Shelves adust in 1 in. increments with patented shelf-release corners.
  • Shelf releases and wedges offer improved rigidity.
  • Assembles quickly without tools; assembled units are nonconductive.
Order Info
Posts sold separately.
Specifications
Product Type Wire Shelf
Dimensions (L x W) 42 x 24 in. (106.6 x 61 cm)
For Use With (Application) Ideal for Drying, Non-corrosive Environments
For Use With (Equipment) Metro™ Super Erecta™ adjustable shelving system
Includes Four non conductive sleeve/wedges
Length (English) 42 in.
Length (Metric) 106.6 cm
Width (English) 24 in.
Width (Metric) 61 cm
Material Chrome