Learn More
Scotch-Brite™ Surface Preparation Pad Plus SPPP17
Product Code. 17257670
Change view
Click to view available options
Quantity:
5/Cs.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17257670 5/Cs. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17257670 Supplier Scotch-Brite™ Supplier No. 7100151294
  • ONLINE EXCLUSIVE PRICE
    178.00€
    91.33€ / 5 pièces
    Économisez 86.67€
    48% de réduction
Expédition estimée: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Surface Preparation Pad Plus
  • Effectively strip and prepare surfaces such as wood, vinyl tile, sheet vinyl, marble, terrazzo and concrete floors
  • Requires only neutral cleaner or water to effectively remove existing floor finishes, preparing the floor for further finishing applications
  • Up to two times faster finish removal than competitive pads when worn
  • Does not require the use of a backer pad (confirm pad driver compatibility with machine manufacturer before using without backer pad)
  • Performs better than competitor pads throughout its service life
  • Helps reduce labour hours and material costs related to stripping floors
  • Does not require the use of stripping chemicals
  • Certified by Green Seal™ for Environmental Innovation for significantly improving mid-life performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Quantity 5/Cs.