Learn More
Pyrex™ SVL™ Stirrer Heads for SVL™ Stirrer Shafts
Product Code. 15537740
Change view
Click to view available options
Type:
Anchor
Collapsible Impeller
Helix - 6 Blades
Paddle
Diameter (Metric):
54 mm
70 mm
80 mm
90 mm
Unit Size:
Each
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Diameter (Metric) Unit Size
15537740 Anchor 54 mm Pack of 5
12024422 Collapsible Impeller 70 mm Each
12044422 Anchor 90 mm Each
12054422 Helix - 6 Blades 80 mm Each
15527740 Paddle 70 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15537740 Supplier Pyrex™ Supplier No. 58898
  • ONLINE EXCLUSIVE PRICE
    500.00€
    376.96€ / 5 pièces
    Économisez 123.04€
    24% de réduction
Expédition estimée: 08-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Switch stirring heads quickly and easily. Pyrex™ SVL™ Stirrer Head for SVL™ Stirrer Shafts are constructed of durable PTFE polymer; some types are fortified with a metal core.
  • Designed for use with SVL™ stirrer shafts
  • Head simply screws into the end of the shaft
  • Manufactured from a PTFE based polymer with the Anchor and Helix types stiffened by a metal core
Specifications
Diameter (Metric) 54 mm
For Use With (Equipment) SVL reaction vessels
Quantity 1 Each
Type Anchor