Learn More
Hettich Lab Technology™ Fixed Angle Rotor
Product Code. 10383670
Change view
Click to view available options
Capacity:
12 x 1.1 to 15 mL
6 x 1.5 to 85 mL
Max. Speed:
6,000 rpm
Max. RCF:
4,146 x g
4,226 x g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Max. Speed Max. RCF unitSize
10383670 6 x 1.5 to 85 mL 6,000 rpm 4,226 x g Each
10191372 12 x 1.1 to 15 mL 6,000 rpm 4,146 x g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10383670 Supplier Hettich Lab Technology™ Supplier No. 1620A
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Fixed Angle Rotor has a relatively low drag for high-speed centrifugation and oblique angling to speed up sedimentation.
  • Used primarily in the higher speed range.
  • Relatively low drag.
  • Oblique angling of tubes shortens the overall distance that particles must travel, thereby speeding up sedimentation.
Specifications
Type Fixed Angle Rotor
Capacity 6 x 1.5 to 85 mL
Max. RCF 4,226 x g
Max. Speed 6,000 rpm
Max. Tube Length 120 mm
Max. Tube Diameter 38 mm
For Use With (Equipment) Universal 320, Rotofix 32A
Angle 35°
No. of Places 6