Learn More
Hettich Lab Technology™ Swing-out Rotor for ROTINA 420/420 R, 4-place
Product Code. 10440374
Change view
Click to view available options
Capacity:
4 x 600 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10440374 4 x 600 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10440374 Supplier Hettich Lab Technology™ Supplier No. 4753
 Restricted Item
Expédition estimée: 06-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Well equipped for all centrifuging applications. ROTINA 420 Centrifuges are ideal for applications in clinical chemistry, in biotechnology and in life science research facilities.
  • Swing-out Rotor
  • 90°, 4-place
  • For use with ROTINA 420/420 R Centrifuge
Specifications
Type Swing-out Rotor
Capacity 4 x 600 mL
Max. RCF 3,095 x g
Max. Speed 4,000 rpm
Radius (Metric) 168 mm
Radius (Metric) Min. 109 mm
Max. Tube Length 100 mm
Max. Tube Diameter 44 mm
For Use With (Equipment) ROTINA 420/420 R
Angle 90°
No. of Places 4