Learn More
Pacific Handy Cutter Swivel Holster for RSC-432
Product Code. 17739047
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17739047 Black Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17739047 Supplier Pacific Handy Cutter Supplier No. UKH443
  • ONLINE EXCLUSIVE PRICE
    107.57€
    92.87€ / 10 pièces
    Économisez 14.70€
    13% de réduction
Expédition estimée: 24-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Practical belt holster made of plastic with fastening clip for belt or clothing. The knife holder can be swivelled.
  • As a result, a comfortable wearing position can be achieved at any time, whether standing or sitting.
  • Suitable for the RSC432 safety knife.
  • Particularly easy to clean.
Specifications
For Use With (Equipment) RSC-432
Material Plastic
Color Black
Sterility Non-sterile