Learn More
Syringol Gentiobioside, TRC
Product Code. 30138312
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138312 2.5 mg 2.5mg
30136011 5 mg 5mg
30007119 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138312 Supplier Toronto Research Chemicals Supplier No. TRCG5969252.5MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1416253-72-9
Molecular Formula C20 H30 O13
Molecular Weight (g/mol) 478.444
Synonym (2S,3R,4S,5S,6R)-2-(2,6-Dimethoxyphenoxy)-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol, Syringol Gentobioside
IUPAC Name (2R,3R,4S,5S,6R)-2-[[(2R,3S,4S,5R,6S)-6-(2,6-dimethoxyphenoxy)-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES COc1cccc(OC)c1O[C@@H]2O[C@H](CO[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C20H30O13/c1-28-8-4-3-5-9(29-2)18(8)33-20-17(27)15(25)13(23)11(32-20)7-30-19-16(26)14(24)12(22)10(6-21)31-19/h3-5,10-17,19-27H,6-7H2,1-2H3/t10-,11-,12-,13-,14+,15+,16-,17-,19-,20+/m1/s1
Formula Weight 478.169
Percent Purity >95%
Chemical Name or Material 2,6-Dimethoxyphenyl-6-O- b-D-glucopyranosyl b-D-Glucopyranoside
Grade HPLC