Learn More
CELLPATH SYSTEM II+ BIOPSY CASSETTE - WHITE LASER (THREADED)
Product Code. 30226253
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 2000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30226253 White Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30226253 Supplier CELLPATH Supplier No. EAM140272B
  • ONLINE EXCLUSIVE PRICE
    436.05€
    245.96€ / 2000 pièces
    Économisez 190.09€
    43% de réduction
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The System II+ Laser Biopsy Cassette comes with a separate white lid that is easily attached via the use of a hinge. Incorporating 0.9 x 0.9 mm apertures, the biopsy cassette offers improved specimen security for small biopsy and tissue fragments.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Product Type Cassette