Learn More
IKA™ T 18 digital Solution
Product Code. 31454120 Shop All IKA Products
Change view
Click to view available options
Plug Type:
CH
EU
UK
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type
31454120 UK
31454108 EU
31454117 CH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 31454120 Supplier IKA™ Supplier No. 0009024702
  • ONLINE EXCLUSIVE PRICE
    2560.00€
    2429.14€ / 1 pièces
    Économisez 130.86€
    5% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
The T 18 digital Solution provides a complete starter setup for homogenization with a dispersing tool, shaft, and stand. This classic, cost-effective version is ideal for heavy-duty, small-scale processes across various industries.
Competitively priced dispersing instrument for volumes from 1 to 1500 mL (H2O) with digital speed display. It offers a wide speed range from 3000 to 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
  • Digital speed display
  • Electronic speed control
  • Electronic overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Plastic disposable dispersing elements are available in two sizes
  • Error code display
  • Quiet operation
Scope of delivery:
  • T 18 digital ULTRA-TURRAX™
  • S 18 N - 19 G Dispersing tool
  • R 1826 Plate stand
  • R 182 Boss head clamp
TRUSTED_SUSTAINABILITY
Specifications
Product Type Homogenizer Package
Speed ≤460 rpm
Voltage 220/240 V
Packaging Type Individual Pack
Plug Type UK