Learn More
Prestige Medical T-Classic Autoclave
Product Code. 15679176
Change view
Click to view available options
Capacity (Metric):
9 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15679176 9 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15679176 Supplier Prestige Medical Supplier No. 210001XP
  • ONLINE EXCLUSIVE PRICE
    1605.53€
    1265.00€ / 1 pièces
    Économisez 340.53€
    21% de réduction
 Restricted Item
Expédition estimée: 23-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
T-Classic, 9 L benchtop autoclave, 126°C cycle, with european plug.
T-Classic, 9 L benchtop autoclave, 126°C cycle, european plug
  • Lightweight and portable
  • 9 liter capacity
  • Sterilizes at 126°C
The Classic is reliable and trusted worldwide, designed to give you the most cost effective sterilization solution. It is simplicity itself to use, fully automatic with a simple, one touch operation and guaranteed to meet your needs.
Specifications
Capacity (English) 2 gal.
Capacity (Metric) 9 L
Depth (Metric) Chamber 230 mm
Diameter (English) Chamber 8.4 in.
Depth (English) Chamber 9.2 in.
Diameter (Metric) Chamber 210 mm
Length (English) 13.6 in.
Length (Metric) 340 mm
Product Type Autoclave
Temperature (English) 250°F to 273°F
Temperature (Metric) 121°C to 134°C
Sterility Non-sterile