Learn More
GVS T-link with Bump Cap and Breathing Tube with Low Flow Indicator
Product Code. 18429655
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
18429655 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429655 Supplier GVS Supplier No. 1712012
  • ONLINE EXCLUSIVE PRICE
    494.00€
    432.82€ / 1 pièces
    Économisez 61.18€
    12% de réduction
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
GVS T-link with bump cap, breathing tube, and low flow indicator.
Accessory
Specifications
Color White
Disposable Reusable
Material Tychem 2000
Product Type Loose Fitting Hood
Size One Size
With Exhalation Valve No