missing translation for 'onlineSavingsMsg'
Learn More

T4 DNA Ligase, in 20mM Tris HCl and 50mM KCl, Thermo Scientific™

Product Code. 15410437
Click to view available options
Quantity:
1 kU
5 kU
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 15410437

Brand: Thermo Scientific Alfa Aesar J65312.4I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kU
Molecular Weight (g/mol) 62 kd
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.