Learn More
Tamoxifen-13C6, TRC
Product Code. 30009553
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009553 1 mg 1mg
30005331 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009553 Supplier Toronto Research Chemicals Supplier No. TRCT0060021MG
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1346606-38-9
Molecular Formula C2013C6H29NO
Molecular Weight (g/mol) 377.47
Synonym (Z)-2-[4-[1-(phenyl-13C6)-2-phenyl-1-butenyl]phenoxy]-N,N-dimethylethanamine, ICI 47699-13C6, Mammaton-13C6, Novaldex-13C6, Tamoxifen-13C6, Z-Tamoxifen-13C6, trans-Tamoxifen-13C6;
SMILES [H][13C]([13CH]=[13CH]1)=[13CH][13CH]=[13C]1/C(C2=CC=C(OCCN(C)C)C=C2)=C(C3=CC=CC=C3)\CC
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C26H29NO/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3/h5-18H,4,19-20H2,1-3H3/b26-25-/i6+1,9+1,10+1,13+1,14+1,22+1
Formula Weight 377.245
Chemical Name or Material Tamoxifen-13C6