Learn More
Tamoxifen citrate, Thermo Scientific™
Inhibitor of protein kinase C and induces apoptosis in cancer cells
202.00€ - 477.00€
Produits 2
Code produit Marque dimQuantity Prix Quantité et disponibilité  
Code produit Marque dimQuantity Prix Quantité et disponibilité  
15413639
FDS Afficher les documents
Thermo Scientific Alfa Aesar
J60955.03
202.00€
1g
Voir les produits de remplacement
Économisez jusqu'à 
»
15423639
FDS Afficher les documents
Thermo Scientific Alfa Aesar
J60955.06
477.00€
5g
Voir les produits de remplacement
Économisez jusqu'à 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Tamoxifen citrate is used as an antiestrogen with potent anti-cancer and PKC inhibitory activities. Tamoxifen Citrate functions as an estrogen receptor antagonist in breast tissue. Tamoxifen Citrate is an agonist towards this receptor in tissues of the endometrium. Tamoxifen Citrate inhibits DNA synthesis and transcription by recruiting co-repressors, which stop estrogen from interacting with genes. When combined with PAX2, Tamoxifen Citrate can inhibit the proliferation of ERBB2. It is also a high affinity activator of the GPR30 receptor. Tamoxifen Citrate is an activator of Estrogen Receptor.
Solubility
Soluble in methanol (50 mg/ml with heat) or ethanol (10 mg/ml with sonication); very slightly soluble in water (0.3 mg/L @ 20°C; the pH is approximately 3.0-3.5), 0.02 N HCl (0.2 mg/ml @ 37°C)4, acetone ,chloroform,and DMSO(100mM)
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.