Learn More
SI Analytics™ Tapper Adapter
Product Code. 10614091 Shop All SI Analytics Products
Change view
Click to view available options
For Use With (Equipment):
12 mm Electrodes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10614091 12 mm Electrodes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10614091 Supplier SI Analytics™ Supplier No. 285215229
  • ONLINE EXCLUSIVE PRICE
    57.80€
    52.00€ / 1 pièces
    Économisez 5.80€
    10% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
SI Analytics™ Tapper Adapter for use with electrodes 12 mm in diameter.
  • NS 14.5
  • PTFE
  • For 12 mm electrodes
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 12 mm Electrodes
Type Tapper Adapter