Learn More
Thermo Scientific™ Temperature Chambers for HAAKE™ MARS™ Rheometers
Shop All Thermo Scientific ProductsDescription
The Thermo Scientific™ HAAKE™ MARS™ rheometer platforms feature temperature chambers which ensure reliable and accurate sample temperature during rheological measurement of melts and solids over a wide temperature range.
Temperature chambers for HAAKE MARS 40/60 and HAAKE MARS iQ Air Rheometers are available.
Both solutions offer advantages such as:
- Patented combination of convection and radiation heat transfer for fast temperature changes and homogeneous temperature distribution
- Fast sample loading, trimming and cleaning due to easy sample access
- Wide portfolio of measuring geometries
- Combination of convection and radiation heat transfer for fast temperature changes and homogeneous temperature distribution from 30°C to 600°C (can be extended to -150°C with the low temperature option)
- CTC for the HAAKE MARS Rheometer platform consists of two halves that allow convenient sample handling and optimal access
- Each half can be moved independently backward and sideways with no special tools required
- When closed, the two halves automatically center themselves around the sample
- Each chamber is equipped with a window to observe the sample during measurement
- A parking position feature allows the attachment of another temperature control unit without removing the CTC
- While in the parked position, the CTC measuring temperature can be set, and the sensors can be cleaned or the next sample can be prepared
- No time is wasted waiting for the CTC to reach its set temperature
- The CTC's temperature range spans from 30°C to 600°C and can be expanded down to -150°C with an optional liquid nitrogen device
Specifications
Specifications
| Type | Clamps |
| Description | Self-centering and self-adjusting clamps for Dynamic Mechanical Thermal Analysis (DMTA) of solid specimen |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.