Learn More
Thermo Scientific™ Temperature Control Modules for the HAAKE™ MARS Rheometer
Product Code. 17690687 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Electrical Temperature Module
Guide Bar Holder
For Use With (Equipment):
HAAKE MARS Active/Passive Hoods
HAAKE MARS Rheometers
HAAKE MARS/MARS iQ Cones and Plates
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
17690687 Electrical Temperature Module HAAKE MARS Rheometers Each
18837244 Electrical Temperature Module HAAKE MARS/MARS iQ Cones and Plates Each
17660687 Guide Bar Holder HAAKE MARS Active/Passive Hoods Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17690687 Supplier Thermo Scientific™ Supplier No. 2222172
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
A sophisticated portfolio of temperature modules covering a broad temperature range is available for Thermo Scientific™ Rheometers.
A sophisticated portfolio of temperature modules is available for Thermo Scientific™ rheometers - covering a broad temperature range. Depending on the application, electrical and liquid controlled temperature modules or Peltier are available. In addition, a controlled temperature test chamber for the high end rheometer platform Thermo Scientific™ HAAKE™ MARS Rheometer is offered.
Benefits of the temperature modules at a glance:
  • Plug-and-play modules with new compact design
  • Automatic temperature module recognition
  • Easy installation in seconds without tools or adjustment
  • Standardized connections including quick connects for cooling media and electronics for all modules
  • Very low thermal mass for fast control response time
  • Integrated base so that the dismounted unit can be parked on the lab bench, for example
TRUSTED_SUSTAINABILITY
Specifications
Type Electrical Temperature Module
Description Electrical temperature module TM-EL-H for HAAKE MARS Rheometers
For Use With (Equipment) HAAKE MARS Rheometers
Required Accessories Holder for guide bar accessory 222-1902
Temperature Range (Metric) -40°C to +400°C