Learn More
TFA Dostmann Temperature/Humidity Transmitter
Product Code. 17754239
Change view
Click to view available options
Type:
Temperature/Humidity Transmitter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17754239 Temperature/Humidity Transmitter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17754239 Supplier TFA Dostmann Supplier No. 30.3187.IT
  • ONLINE EXCLUSIVE PRICE
    20.02€
    16.14€ / 1 pièces
    Économisez 3.88€
    19% de réduction
Expédition estimée: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Transmitter for TFA wireless thermo-hygrometers and weather stations.
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3044, 30.3045, 35.1109, 35.1111, 35.1113, 35.1121
  • For wireless transmission of the outdoor climate
  • Display with temperature and humidity
  • Range: 100 m in open field
  • Update: every 4 seconds
  • Also for climate monitoring of remote rooms
Specifications
Type Temperature/Humidity Transmitter