Learn More
TFA Dostmann Temperature Transmitter
Product Code. 16671246
Change view
Click to view available options
Type:
Temperature/Humidity Transmitter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
16671246 Temperature/Humidity Transmitter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 16671246 Supplier TFA Dostmann Supplier No. 30.3229.02
  • ONLINE EXCLUSIVE PRICE
    18.16€
    13.32€ / 1 pièces
    Économisez 4.84€
    26% de réduction
Expédition estimée: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Transmitter for TFA wireless thermo-hygrometers and weather stations.
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3054, 30.3056, 30.3062
  • For wireless transmission of temperature
  • With waterproof cable sensor
  • For fridge, freezer, aquarium, pool etc.
  • Also for temperature monitoring in remote rooms
Specifications
Type Temperature/Humidity Transmitter