Learn More
tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate, 95%, Thermo Scientific™
Product Code. 15229696 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
Product Code. 15229696
Brand: Thermo Scientific Acros 451232500
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products
This item is not returnable. View return policy
Chemical Identifiers
CAS 898271-20-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
InChI Key XQNQFOKZUQWWDO-UHFFFAOYSA-N
Synonym 1-boc-3-aminoethylazetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, 1-boc-3-aminoethyl azetidine, 1-n-boc-3-aminoethyl azetidine, 1-boc-3-2-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate
PubChem CID 44630603
IUPAC Name tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)CCN
Specifications
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 200.28
Percent Purity 95%
Chemical Name or Material tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate
compliance-icons
Product Identifier
  • tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.