Learn More
WTW™ TetraCon™ 925-P Conductivity Electrode
Product Code. 15144155
Change view
Click to view available options
Temperature (Metric) Operating:
0°C to 100°C
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Operating Unit Size
15144155 0°C to 100°C 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15144155 Supplier WTW™ Supplier No. 301716
  • ONLINE EXCLUSIVE PRICE
    918.02€
    660.00€ / 1 pièces
    Économisez 258.02€
    28% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Universal digital conductiviy measurement in aqueous samples. For use with MPP IDS or to connect to the cable AS/IDS-x.
WTW has been manufacturing conductivity measuring cells of the highest quality for decades. These are also the basis for the IDS conductivity measuring cells. The advantages of the IDS technology include the automatic transfer of the cell constant and therefore the exclusion of possible erroneous input.
There are two models available to cover the entire conductivity range:
The four electrode measuring cell TetraCon™ 925 for medium and high conductivity, for pure water and for low conductivity; and the two electrode measuring cell LR 925/01 including flow container. WTW's proven IDS conductivity measuring cells are now available as sustainable plug head versions.
The universal plug head fits the sensor with wireless functionality – disturbing cables are no longer required. Furthermore it can be connected to AS/IDS-x cables with lengths of up to 100 m. With this new technology WTW significantly expands the range of applications and the measuring comfort of its digital conductivity measuring systems.
  • Proven electrode technology
  • Automatic transfer of cell constant and temperature compensation
  • Wide area of application - 1 μS/cm to 2000 mS/cm
  • 4-electrode-IDS conductivity cell with graphite electrodes, epoxy shaft, cell constant 0.475 cm–1, waterproof up to 10 bar (IP 68)
TRUSTED_SUSTAINABILITY
Specifications
Description TetraCon 925-P
For Use With (Equipment) MPP IDS or to Connect to the Cable AS/IDS-x
Type Conductivity Electrode
Includes Graphite four electrode measuring cell, Plug head for AS/IDS-x cables, MPP 930 or wireless modules IDS WLM-S. (Cable is not included.)
Material Epoxy
Temperature (Metric) Operating 0°C to 100°C
Length (Metric) 120 mm
Product Title
Select an issue
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.