Learn More
Tetrahydro-1,3,4,6-tetramethylimidazo(4,5-d)imidazole-2,5(1H,3H)-dione, TRC
Product Code. 30452401
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452401 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452401 Supplier Toronto Research Chemicals Supplier No. TRCT889843200MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mebicar
CAS 10095-06-4
Molecular Formula C8 H14 N4 O2
Quantity 200 mg
Synonym Imidazo[4,5-d]imidazole-2,5(1H,3H)-dione','tetrahydro-1,3,4,6-tetramethyl-, Glycoluril','1,3,4,6-tetramethyl- (7CI), Tetrahydro-1,3,4,6-tetramethylimidazo[4,5-d]imidazole-2,5(1H,3H)-dione, Mebicar, Mebikar
InChI Formula InChI=1S/C8H14N4O2/c1-9-5-6(11(3)7(9)13)12(4)8(14)10(5)2/h5-6H,1-4H3
SMILES CN1C2C(N(C)C(=O)N2C)N(C)C1=O
IUPAC Name 1,3,4,6-tetramethyl-3a,6a-dihydroimidazo[4,5-d]imidazole-2,5-dione
Molecular Weight (g/mol) 198.22
Formula Weight 198.1117