Learn More
Tetrahydrofuran D8 99.50% D, Eurisotop
Product Code. 17297436
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
2 x 0.75 mL
2 x 1 mL
25 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17297436 2 x 0.75 mL
17207446 2 x 1 mL
17227446 10 mL
17217446 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17297436 Supplier Eurisotop Supplier No. D149BB
 Restricted Item
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Synonym: Tetrahydrofuran-d8, Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran
Tetrahydrofuran D8
  • Isotopic Enrichment: 99.50% D
CoA and MSDS available at Eurisotop.com
Chemical Identifiers
CAS 1693-74-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 80.16
InChI Key WYURNTSHIVDZCO-SVYQBANQSA-N
Synonym Tetrahydrofuran-d8, Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran
IUPAC Name (2H8)oxolane
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
Specifications
Color Colorless
Deuterium (D) 99.50% D
Packaging Ampule
Quantity 2 x 0.75 mL
Melting Point -108.5°C
Boiling Point 66°C
Grade Research
Chemical Name or Material Tetrahydrofuran D8