Learn More
Tetrathiafulvalene - 7,7,8,8-Tetracyanoquinodimethane Complex, TRC
Product Code. 30456434
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456434 100 mg 100mg
30453916 10 mg 10mg
30450673 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456434 Supplier Toronto Research Chemicals Supplier No. TRCT310965100MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40210-84-2
Molecular Formula C6H4S4 . C12H4N4
Molecular Weight (g/mol) 408.54
IUPAC Name 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile;2-(1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES N#CC(=C1C=CC(=C(C#N)C#N)C=C1)C#N.S2C=CSC2=C3SC=CS3
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C12H4N4.C6H4S4/c13-5-11(6-14)9-1-2-10(4-3-9)12(7-15)8-16;1-2-8-5(7-1)6-9-3-4-10-6/h1-4H;1-4H
Formula Weight 407.9632
Chemical Name or Material Tetrathiafulvalene - 7,7,8,8-Tetracyanoquinodimethane Complex