Learn More
Thermo Scientific™ Barnstead™ Deionization System
Shop All Thermo Scientific Products
Click to view available options
Type:
Deionization Cartridge
Deionization System
Unit Size:
Each
Description
Deionization System
For the Nanopure, E-pure Water Systems
Specifications
Specifications
| Pore Size | 60 μm |
| Resin Type | Mixed Bed |
| For Use With (Application) | Ultrapure™ ion exchange |
| For Use With (Equipment) | Nanopure Water System, Ultrapure Ion Exchange |
| Type | Deionization System |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction