Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
Product Code. 10384681 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Green
Light Gray
Olive Brown
Red
White
Yellow
Tube Size:
10/15 mL
15 mL
175/250 mL
50 mL
7 mL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Tube Size Unit Size
10384681 Olive Brown 15 mL Each
10290162 Yellow 7 mL Each
10782393 Light Gray 7 mL Each
10202991 Red 15 mL Each
10259301 White 10/15 mL Each
10395472 Green 50 mL Each
10181031 White 175/250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10384681 Supplier Thermo Scientific™ Supplier No. 75008137
  • ONLINE EXCLUSIVE PRICE
    182.00€
    141.81€ / 1 pièces
    Économisez 40.19€
    22% de réduction
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441
Adapters sold individually; order correct quantity to equip your rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LH-4000W and LH-4000 rotor with round bucket
Color Olive Brown
Tube Size 15 mL