Learn More
Sonoco™ ThermoSafe LabMailer Specimen Tube Mailers
Product Code. 17808703
Change view
Click to view available options
Capacity:
7, 10, 15 or 25 mL
Quantity:
5 Tubes
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Quantity Unit Size
17808703 7, 10, 15 or 25 mL 5 Tubes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17808703 Supplier Sonoco™ Supplier No. 409
  • ONLINE EXCLUSIVE PRICE
    232.47€
    133.07€ / 100 pièces
    Économisez 99.40€
    42% de réduction
View all promotions
Expédition estimée: 22-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Shock-absorbing polystyrene foam mailers accept sample tubes with rubber or screw caps
  • Two-piece assembly with snap-lock fasteners holds contents in place
  • Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples.
†Includes slotted area for up to four slides.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 Tubes
Applications Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples
Type EPS Shipper
Dimensions (L x W x H) Exterior 7.875 x 5.125 x 1.5 in.
Width (Metric) Interior 16 mm
Diameter (Metric) Interior 170 mm
Description Foam only shipper. Must purchase mailing sleeve model 410, Holds 5 Tubes
Capacity 7, 10, 15 or 25 mL
Material Polystyrene Foam