missing translation for 'onlineSavingsMsg'
Learn More

Thiamine nitrate

Product Code. 15472859
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 15472859

Brand: Thermo Scientific Alfa Aesar J60014.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 532-43-4 | C12H17N5O4S | 327.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-43-4
Molecular Formula C12H17N5O4S
Molecular Weight (g/mol) 327.36
MDL Number MFCD00036330
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
Synonym thiamine nitrate, thiamine mononitrate, vitamin b1 nitrate, vitamin b1 mononitrate, aneurine mononitrate, aneurine nitrate, betabion mononitrate, thiamine nitrate salt, vitamin b sub 1 nitrate, thiamine mononitrate usp
PubChem CID 10762
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;nitrate
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

Melting Point 190°C
Color White
Quantity 10 g
Merck Index 14,9295
Formula Weight 327.36
Physical Form Powder
Chemical Name or Material Thiamine nitrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.